Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-{3-[(tert-butoxycarbonyl)amino]-1-pyrrolidinyl}-5-nitronicotinate ≥95%

Supplier: MATRIX SCIENTIFIC MS

056358-500MG 056358-1G
101910-184EA 556.92 USD InStock
101910-184 101910-186
Methyl-2-{3-[(tert-butoxycarbonyl)amino]-1-pyrrolidinyl}-5-nitronicotinate ≥95%
Methyl-2-{3-[(tert-butoxycarbonyl)amino]-1-pyrrolidinyl}-5-nitronicotinate
Hazard: Irritant


Formula: C₁₆H₂₂N₄O₆
MDL Number: MFCD16622812
CAS Number: 1242268-00-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty