Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9H-Fluoren-9-ylmethyl 3-(aminomethyl)piperidine-1-carboxylate hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

056407-500MG 056407-1G
101910-384EA 375.38 USD InStock
101910-384 101910-386
9H-Fluoren-9-ylmethyl 3-(aminomethyl)piperidine-1-carboxylate hydrochloride ≥95%
9H-Fluoren-9-ylmethyl 3-(aminomethyl)piperidine-1-carboxylate hydrochloride
Hazard: Irritant


Formula: C₂₁H₂₅ClN₂O₂
MW: 372.9 g/mol
MDL Number: MFCD03001694
CAS Number: 669713-56-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty