Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichlorobenzyl thiocyanate ≥98%

Supplier: MATRIX SCIENTIFIC MS

057006-10G 057006-50G
101912-018EA 73.85 USD InStock
101912-018 101912-020
2,6-Dichlorobenzyl thiocyanate ≥98%
2,6-Dichlorobenzyl thiocyanate
Hazard: Irritant-Harmful


Formula: C₈H₅Cl₂NS
MW: 218.11 g/mol
MDL Number: MFCD00019766
CAS Number: 7534-64-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty