Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethoxybenzyl thiocyanate ≥98%

Supplier: MATRIX SCIENTIFIC MS

057027-10G 057027-50G
101912-096EA 156.92 USD InStock
101912-096 101912-098
2,5-Dimethoxybenzyl thiocyanate ≥98%
2,5-Dimethoxybenzyl thiocyanate
Hazard: Irritant


Formula: C₁₀H₁₁NO₂S
MW: 209.27 g/mol
MDL Number: MFCD09025688
CAS Number: 1252046-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty