Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,4-Difluorophenyl)acetohydrazide ≥95%

Supplier: MATRIX SCIENTIFIC MS

058224-500MG 058224-1G 058224-5G
101914-290EA 278.46 USD InStock
101914-290 101914-292 101914-294
2-(2,4-Difluorophenyl)acetohydrazide ≥95%
2-(2,4-Difluorophenyl)acetohydrazide
Hazard: Irritant


Formula: C₈H₈F₂N₂O
Melting Pt: 145…147 °C
MDL Number: MFCD17079088
CAS Number: 925252-63-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty