Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(1H-Tetrazol-5-yl)thiophene-2-sulfonyl chloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

068402-500MG 068402-1G
101935-976EA 1512.31 USD InStock
101935-976 101935-978
5-(1H-Tetrazol-5-yl)thiophene-2-sulfonyl chloride ≥95%
5-(1H-Tetrazol-5-yl)thiophene-2-sulfonyl chloride
Hazard: Irritant


Formula: C₅H₃ClN₄O₂S₂
MW: 250.69 g/mol
MDL Number: MFCD09263262
CAS Number: 924964-21-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty