Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoxazole-3-carbaldehyde ≥95%

Supplier: MATRIX SCIENTIFIC MS

068500-250MG 068500-500MG
101936-412EA 398.46 USD InStock
101936-412 101936-414
Isoxazole-3-carbaldehyde ≥95%
Isoxazole-3-carbaldehyde
Hazard: Irritant


Formula: C₄H₃NO₂
MDL Number: MFCD06739019
CAS Number: 89180-61-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty