Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indole-6-carboxylic acid, 2,3-dihydro-2,3-dioxo-, methyl ester ≥95%

Supplier: MATRIX SCIENTIFIC MS

068514-250MG 068514-500MG
101936-478EA 844.62 USD InStock
101936-478 101936-480
1H-Indole-6-carboxylic acid, 2,3-dihydro-2,3-dioxo-, methyl ester ≥95%
1H-Indole-6-carboxylic acid, 2,3-dihydro-2,3-dioxo-, methyl ester
Hazard: Irritant


Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
MDL Number: MFCD17214341
CAS Number: 213670-35-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty