Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(tert-Butoxycarbonyl)-3-iodopyrrolidine ≥95%

Supplier: MATRIX SCIENTIFIC MS

068627-250MG 068627-500MG
101936-982EA 669.23 USD InStock
101936-982 101936-984
1-(tert-Butoxycarbonyl)-3-iodopyrrolidine ≥95%
1-(tert-Butoxycarbonyl)-3-iodopyrrolidine
Hazard: Irritant


Formula: C₉H₁₆INO₂
MW: 297.14 g/mol
MDL Number: MFCD12031283
CAS Number: 774234-25-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty