Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (S)-piperidine-2-carboxylate hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

068950-1G 068950-5G 068950-25G
101938-408EA 36.92 USD InStock
101938-408 102551-728 102551-730
Methyl (S)-piperidine-2-carboxylate hydrochloride ≥95%
Methyl (S)-piperidine-2-carboxylate hydrochloride
Hazard: Irritant


Formula: C₇H₁₄ClNO₂
MW: 179.6446 g/mol
MDL Number: MFCD00672366
CAS Number: 18650-39-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty