Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-10-benzyl-11-oxo-4,10-diazaspiro[5.5]undecane-4-carboxylate ≥95%

Supplier: MATRIX SCIENTIFIC MS

069018-250MG 069018-500MG 069018-1G
101938-648EA 698.46 USD InStock
101938-648 101938-650 101938-652
tert-Butyl-10-benzyl-11-oxo-4,10-diazaspiro[5.5]undecane-4-carboxylate ≥95%
tert-Butyl-10-benzyl-11-oxo-4,10-diazaspiro[5.5]undecane-4-carboxylate
Hazard: Irritant


Formula: C₂₁H₃₀N₂O₃
MDL Number: MFCD12912610
CAS Number: 1198286-10-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty