Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Methylthiazol-5-yl)ethylamine ≥97%

Supplier: MATRIX SCIENTIFIC MS

069076-500MG 069076-1G 069076-5G
101938-820EA 466.15 USD InStock
101938-820 101938-822 101938-824
2-(4-Methylthiazol-5-yl)ethylamine ≥97%
2-(4-Methylthiazol-5-yl)ethylamine
Hazard: Irritant


Formula: C₆H₁₀N₂S
MDL Number: MFCD08060722
CAS Number: 58981-35-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty