Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7,8-Diamino-2-(4-(dimethylamino)phenyl)-4H-chromen-4-one trihydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

069127-250MG 069127-500MG
101939-036EA 1221.54 USD InStock
101939-036 101939-038
7,8-Diamino-2-(4-(dimethylamino)phenyl)-4H-chromen-4-one trihydrochloride ≥95%
7,8-Diamino-2-(4-(dimethylamino)phenyl)-4H-chromen-4-one trihydrochloride
Hazard: Irritant


Formula: C₁₇H₂₀Cl₃N₃O₂
MDL Number: MFCD18433529
CAS Number: 1258638-65-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty