Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,2-Trimethylpiperazine hydrochloride ≥95%

Supplier: MATRIX SCIENTIFIC MS

069184-250MG 069184-500MG
101939-224EA 756.92 USD InStock
101939-224 101939-226
1,2,2-Trimethylpiperazine hydrochloride ≥95%
1,2,2-Trimethylpiperazine hydrochloride
Hazard: Irritant


Formula: C₇H₁₇ClN₂
MDL Number: MFCD09864551
CAS Number: 932047-03-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty