Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-(chloromethyl)thiazole ≥97%

Supplier: MATRIX SCIENTIFIC MS

069213-25G 069213-100G
101939-300EA 33.85 USD InStock
101939-300 102551-742
2-Chloro-5-(chloromethyl)thiazole ≥97%
2-Chloro-5-(chloromethyl)thiazole
Hazard: Toxic/Corrosive/Refrigerate


Formula: C₄H₃Cl₂NS
Melting Pt: 29…31 °C
MDL Number: MFCD01073549
CAS Number: 105827-91-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty