Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,4S)-tert-Butyl-3-oxo-2-oxa-5-azabicyclo-[2.2.1]heptane-5-carboxylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

070429-1G 070429-5G 070429-25G
101944-714EA 553.85 USD InStock
101944-714 101944-716 101944-718
(1S,4S)-tert-Butyl-3-oxo-2-oxa-5-azabicyclo-[2.2.1]heptane-5-carboxylate ≥97%
(1S,4S)-tert-Butyl-3-oxo-2-oxa-5-azabicyclo-[2.2.1]heptane-5-carboxylate
Hazard: Irritant


Formula: C₁₀H₁₅NO₄
MW: 213.24 g/mol
MDL Number: MFCD06797016
CAS Number: 113775-22-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty