Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyridin-3-yl-oxoacetic acid ethyl ester ≥96%

Supplier: MATRIX SCIENTIFIC MS

070449-500MG 070449-1G 070449-5G
101944-830EA 930.77 USD InStock
101944-830 101944-832 101944-834
Imidazo[1,2-a]pyridin-3-yl-oxoacetic acid ethyl ester ≥96%
Imidazo[1,2-a]pyridin-3-yl-oxoacetic acid ethyl ester
Hazard: Irritant


Formula: C₁₁H₁₀N₂O₃
MW: 218.21 g/mol
MDL Number: MFCD13183153
CAS Number: 603301-58-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty