Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethenylazetidine-1-carboxylic acid tert-butyl ester ≥95%

Supplier: MATRIX SCIENTIFIC MS

070635-250MG 070635-1G
101945-674EA 2093.85 USD InStock
101945-674 101945-676
3-Ethenylazetidine-1-carboxylic acid tert-butyl ester ≥95%
3-Ethenylazetidine-1-carboxylic acid tert-butyl ester
Hazard: Irritant


Formula: C₁₀H₁₇NO₂
MW: 183.25 g/mol
MDL Number: MFCD16140212
CAS Number: 1026796-78-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty