Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Azaspiro[3.3]heptane-2,6-dicarboxylic acid-2-benzyl ester ≥95%

Supplier: MATRIX SCIENTIFIC MS

070654-500MG 070654-1G
101945-760EA 2210.77 USD InStock
101945-760 101945-762
2-Azaspiro[3.3]heptane-2,6-dicarboxylic acid-2-benzyl ester ≥95%
2-Azaspiro[3.3]heptane-2,6-dicarboxylic acid-2-benzyl ester
Hazard: Irritant


Formula: C₁₅H₁₇NO₄
MW: 275.31 g/mol
MDL Number: MFCD18432778
CAS Number: 1291487-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty