Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester ≥95%

Supplier: MATRIX SCIENTIFIC MS

070655-100MG 070655-250MG 070655-500MG
101945-764EA 1221.54 USD InStock
101945-764 101945-766 101945-768
(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester ≥95%
(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester
Hazard: Irritant


Formula: C₁₂H₁₉NO₃
MW: 225.29 g/mol
MDL Number: MFCD11858160
CAS Number: 1118786-86-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty