Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Oxetanemethanamine ≥95%

Supplier: MATRIX SCIENTIFIC MS

070690-100MG 070690-250MG 070690-500MG
101945-918EA 643.38 USD InStock
101945-918 101945-920 101945-922
2-Oxetanemethanamine ≥95%
2-Oxetanemethanamine
Hazard: Irritant


Formula: C₄H₉NO
MDL Number: MFCD17214171
CAS Number: 882402-12-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty