Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexahydro-1,4-thiazepine hydrochloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

070722-250MG 070722-500MG 070722-1G
101946-056EA 1047.69 USD InStock
101946-056 101946-058 101946-060
Hexahydro-1,4-thiazepine hydrochloride ≥97%
Hexahydro-1,4-thiazepine hydrochloride
Hazard: Irritant


Formula: C₅H₁₂ClNS
MDL Number: MFCD11111967
CAS Number: 108724-14-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty