4CN Membrane Peroxidase Substrate, KPL
Supplier: MetaSystems Hard&Software GmbH
5420-0024 5420-0020
95059-150EA 1112.28 USD InStock
95059-150 95059-148
4CN Membrane Peroxidase Substrate, KPL
Electrophoresis Reagents Western Blotting Reagents
4CN (4-chloro-1-napthol) is a reliable substrate for detection of membrane-bound proteins
4CN provides an ideal alternative to other, more sensitive substrates where background is a problem. It is provided in a convenient, 2-component format, mix-and-use format.
4CN produces a dark purple precipitate at the reaction site in the presence of peroxidase-labeled conjugates. It is recommended for membrane assays and dot blots.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty