Ace-Thred Connectors, Glass, Ace Glass Incorporated
Supplier: Ace Glass
7644-10 7644-15 7644-18 7644-20 7644-25 7644-36
89059-946EA 37 USD InStock
89059-946 89059-948 89059-950 89059-952 89059-954 89059-956 89059-958
Ace-Thred Connectors, Glass, Ace Glass Incorporated
Fittings and Connectors
These internally threaded glass connectors can be used in place of ground glass joints on flasks, columns, etc
Threads offer positive pressure o-ring seal when used with the proper fittings. Fabricated from borosilicate glass. O.D value represents the value of "Thread O.D".
Información para pedidos: Fittings and bushings for all sizes must be ordered separately. For #7 Ace-Thred size, order 89051-092.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty