BetaMax™-ES Liquid Scintillation Cocktail, MP Biomedicals
Supplier: E. Becker & Co. GmbH
0188002004 0188002001 0188002005
IC-88002004CS 789.04 USD InStock
IC-88002004 IC88002001 IC88002005
BetaMax™-ES Liquid Scintillation Cocktail, MP Biomedicals
Scintillation Cocktails
BetaMax™ ES, as its name implies, is a very efficient, environmentally safe LSC cocktail for beta particle counting in non-aqueous samples
  • Biodegradable
  • Non-toxic, Non-flammable & Odorless
  • High Tritium Efficiency
  • Excellent With Dry Filters & Test Wipes
BetaMax™ ES is an environmentally safe, biodegradable, non-toxic, and non-flammable product designed to work well with most non-aqueous samples, samples dried on solid supports, and organic samples extracted from aqueous mixtures.
It is excellent for samples completely dried that have been recovered on filters of all types. Some membrane filters can be dissolved in 1-2 mL of methyl cellosolve which can be mixed with 10-15 mL of BetaMax™ ES for optimum recovery. Free lipids and steroids can be extracted from an aqueous sample by simply adding the aqueous sample to the BetaMax™ ES and allowing the organic soluble components to migrate across the partition. This usually occurs in approximately 30 minutes at room temperature (the time required to adapt samples to darkness).
Primary Applications: Organic samples, dry filters, test wipes and 3H Acetyl CoA CAT assay extractions.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty