Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Naphtholbenzeine 1% (w/v) in titration solvent indicator

Supplier: RICCA CHEMICAL CO
Danger

R5225000-120C R5225000-1C
RC5225-4EA 88 USD InStock
RC5225-4 RC5225-32
1-Naphtholbenzeine 1% (w/v) in titration solvent indicator
1-Naphtholbenzeine


Formula: C₂₇H₁₈O₂
MW: 374.44 g/mol
Boiling Pt: ~ 90 °C
Density: 0.8200 g/cm³
Flash Pt: < 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00078492
CAS Number: 145-50-6
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Appearance Dark red, Toluene odor
Suitability for Use Yellow - Orange to Green/Green-Brown (Acidic) - Green/Brown to Orange (Basic)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty