Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenol red USP Test Solution (TS) pH 4.7

Supplier: RICCA CHEMICAL CO
Danger

R5721000-120A R5721000-500A
RC5721-4EA 51 USD InStock
RC5721-4 RC5721-16
Phenol red USP Test Solution (TS) pH 4.7
Phenol red


Formula: C₁₉H₁₄O₅S
MW: 354.38 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8
Merck Index: 13,07329
Order Now
Specification Test Results
Appearance Clear, yellow, slight Acetate odor
pH at 25 °C (traceable to NIST SRM 185 & 186) 4.70 ± 0.05 pH at 25.0 °C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty