Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Water, high purity for rinsing electrodes and other laboratory instruments

Supplier: RICCA CHEMICAL CO

R6648000-200PK R6648000-500A R6648000-20PK
RC6648-20BEA 688 USD InStock
RC6648-20B RC6648-16 RC6648-20
Water, high purity for rinsing electrodes and other laboratory instruments
Water


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty