A0514-5G
A0514-25G
TCA0514-005GEA
48
USD
InStock
TCA0514-005G
TCA0514-25G
DL-Arabinose ≥98.0%
DL-Arabinose
CAS Number: 147-81-9
MDL Number: MFCD00135867
Molecular Formula: C5H10O5
Molecular Weight: 150.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 158