Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylbutyl acetate ≥98.0%

Supplier: Decon Laboratories Ltd
Danger

A1076-500ML A1076-25ML
TCA1076-500MLEA 165 USD InStock
TCA1076-500ML TCA1076-025ML
2-Methylbutyl acetate ≥98.0%
2-Methylbutyl acetate
CAS Number: 624-41-9
MDL Number: MFCD00040494
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Flash Point (°C): 36
Specific Gravity (20/20): 0.88

Other Product Names / Synonyms: Acetic Acid 2-Methylbutyl Ester


Formula: C₇H₁₄O₂
Boiling Pt: 141 °C
Density: 0.88 (20 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00040494
CAS Number: 624-41-9
EINECS: 210-843-8
UN: 1104
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba9882722d5e3f2f94a7eb9c8a5bfe08c5feeeb&vid=4ba9882722d5e3f2f94a7eb9c8a5bfe08c5feeeb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty