Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromophthalic anhydride ≥97.0%

Supplier: Decon Laboratories Ltd

B1693-25G B1693-500G
TCB1693-025GEA 94 USD InStock
TCB1693-025G TCB1693-500G
4-Bromophthalic anhydride ≥97.0%
4-Bromophthalic anhydride
CAS Number: 86-90-8
MDL Number: MFCD00191323
Molecular Formula: C8H3BrO3
Molecular Weight: 227.01
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 301
Melting point (°C): 107


Formula: C₈H₃BrO₃
Boiling Pt: 301 °C
Melting Pt: 107 °C
MDL Number: MFCD00191323
CAS Number: 86-90-8
EINECS: 201-707-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1786109830a00446675a1394f6fe74a9be7bd7cb&vid=1786109830a00446675a1394f6fe74a9be7bd7cb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty