B1891-500ML
B1891-25ML
TCB1891-500MLEA
178
USD
InStock
TCB1891-500ML
TCB1891-025ML
Hexyl butyrate ≥98.0%
Hexyl butyrate
CAS Number: 2639-63-6
MDL Number: MFCD00048884
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 81
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: Butyric Acid Hexyl Ester