Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chlorobenzotriazole ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

C0137-100G C0137-25G
TCC0137-100GEA 163 USD InStock
TCC0137-100G TCC0137-025G
5-Chlorobenzotriazole ≥98.0% (by GC, titration analysis)
5-Chlorobenzotriazole
CAS Number: 94-97-3
MDL Number: MFCD00005700
Molecular Formula: C6H4ClN3
Molecular Weight: 153.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 158


Formula: C₆H₄ClN₃
Melting Pt: 158 °C
MDL Number: MFCD00005700
CAS Number: 94-97-3
EINECS: 202-378-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff901bb5622aa5d7206bf0f52a3611efbf7d6f6e&vid=ff901bb5622aa5d7206bf0f52a3611efbf7d6f6e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty