Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

m-Cymene ≥99.0%

Supplier: Decon Laboratories Ltd

C0798-1ML C0798-0.1ML
TCC0798-001MLEA 326 USD InStock
TCC0798-001ML TCC0798-0.1ML
m-Cymene ≥99.0%
m-Cymene
CAS Number: 535-77-3
MDL Number: MFCD00008891
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 47
Specific Gravity (20/20): 0.86

Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: 3-Isopropyltoluene, 3-Methylisopropylbenzene


Formula: C₁₀H₁₄
Boiling Pt: 175 °C
Melting Pt: -63.8 °C
Density: 0.86 (20 °C)
Flash Pt: 47 °C
MDL Number: MFCD00008891
CAS Number: 535-77-3
EINECS: 208-617-9
UN: 2046
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efb33649c957a6634261eafdf1ea02e1cc5c82c7&vid=efb33649c957a6634261eafdf1ea02e1cc5c82c7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty