Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorohexane ≥98.0%

Supplier: Decon Laboratories Ltd

C0890-5G C0890-25G
TCC0890-005GEA 142 USD InStock
TCC0890-005G TCC0890-025G
2-Chlorohexane ≥98.0%
2-Chlorohexane
CAS Number: 638-28-8
MDL Number: MFCD00039351
Molecular Formula: C6H13Cl
Molecular Weight: 120.62
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Specific Gravity (20/20): 0.87
Storage Temperature: 0-10°C

Other Product Names / Synonyms: 2-Hexyl Chloride


Formula: C₆H₁₃Cl
Boiling Pt: 122 °C
Density: 0.87 (20 °C)
MDL Number: MFCD00039351
CAS Number: 638-28-8
EINECS: 211-329-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b560ab5888b8d18a65b70f6e1a5c82eb69026cd1&vid=b560ab5888b8d18a65b70f6e1a5c82eb69026cd1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty