Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorophthalic anhydride ≥98.0%

Supplier: Decon Laboratories Ltd

C1328-5G C1328-25G
TCC1328-005GEA 116.1 USD InStock
TCC1328-005G TCC1328-025G
4-Chlorophthalic anhydride ≥98.0%
4-Chlorophthalic anhydride
CAS Number: 118-45-6
MDL Number: MFCD00152354
Molecular Formula: C8H3ClO3
Molecular Weight: 182.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 290
Melting point (°C): 96
Flash Point (°C): 152


Formula: C₈H₃ClO₃
MW: 182.56 g/mol
Boiling Pt: 290 °C
Melting Pt: 96 °C
Flash Pt: 152 °C
MDL Number: MFCD00152354
CAS Number: 118-45-6
EINECS: 204-251-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1483f3ce88b2f70d4559421d62de02a51a9cb4d0&vid=1483f3ce88b2f70d4559421d62de02a51a9cb4d0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty