Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,10-Dichlorodecane ≥97.0%

Supplier: Decon Laboratories Ltd

D0247-25G D0247-500G
TCD0247-025GEA 61.03 USD InStock
TCD0247-025G TCD0247-500G
1,10-Dichlorodecane ≥97.0%
1,10-Dichlorodecane
CAS Number: 2162-98-3
MDL Number: MFCD00000957
Molecular Formula: C10H20Cl2
Molecular Weight: 211.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 146
Freezing point (°C): 14
Specific Gravity (20/20): 1.00

Other Product Names / Synonyms: 1,10-Decamethylene Dichloride


Formula: ClH₂C(CH₂)₈CH₂Cl
MW: 211.17 g/mol
Boiling Pt: 102 °C
Melting Pt: 14…16 °C
Density: 1.00 (20 °C)
Flash Pt: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD00000957
CAS Number: 2162-98-3
EINECS: 218-489-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0505841c9f24fd3381adfa0217b27ef58ef20086&vid=0505841c9f24fd3381adfa0217b27ef58ef20086&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty