Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethylformamide ≥99.5%

Supplier: Decon Laboratories Ltd
Danger

D0722-500ML D0722-25ML D0722-100ML
TCD0722-500MLEA 37.26 USD InStock
TCD0722-500ML TCD0722-25ML TCD0722-100ML
N,N-Dimethylformamide ≥99.5%
N,N-Dimethylformamide
CAS Number: 68-12-2
MDL Number: MFCD00003284
Molecular Formula: C3H7NO
Molecular Weight: 73.10
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 153
Melting point (°C): -61
Flash Point (°C): 60
Specific Gravity (20/20): 0.95

Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: Formyldimethylamine, DMF


Formula: HCON(CH₃)₂
Boiling Pt: 153 °C
Melting Pt: –61 °C
Density: 0.95 (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de9eebe8744933790015429a33188a2f4d7881fa&vid=de9eebe8744933790015429a33188a2f4d7881fa&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.5% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty