Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Decyloxybenzaldehyde ≥97.0%

Supplier: Decon Laboratories Ltd

D1124-5G D1124-25G
TCD1124-5GEA 21 USD InStock
TCD1124-5G TCD1124-025G
4-Decyloxybenzaldehyde ≥97.0%
4-Decyloxybenzaldehyde
CAS Number: 24083-16-7
MDL Number: MFCD00021072
Molecular Formula: C17H26O2
Molecular Weight: 262.39
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 176
Specific Gravity (20/20): 0.96


Formula: C₁₇H₂₆O₂
Boiling Pt: 176 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00021072
CAS Number: 24083-16-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20f9903f13dfe0493e23ffb6808c735cdcd258aa&vid=20f9903f13dfe0493e23ffb6808c735cdcd258aa&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty