Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dihydro-2-methoxy-2H-pyran ≥97.0%

Supplier: Decon Laboratories Ltd

D1554-25ML D1554-500ML
TCD1554-025MLEA 21 USD InStock
TCD1554-025ML TCD1554-500ML
3,4-Dihydro-2-methoxy-2H-pyran ≥97.0%
3,4-Dihydro-2-methoxy-2H-pyran
CAS Number: 4454-05-1
MDL Number: MFCD00006560
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Flash Point (°C): 16
Specific Gravity (20/20): 1.01

Other Product Names / Synonyms: 2-Methoxy-3,4-dihydro-2H-pyran


Formula: C₆H₁₀O₂
Boiling Pt: 127 °C
Density: 1.01 (20 °C)
Flash Pt: 16 °C
MDL Number: MFCD00006560
CAS Number: 4454-05-1
EINECS: 224-698-3
UN: 3271
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=065ec5aa6ffe57e08d870fd1b49a9e8b42212f89&vid=065ec5aa6ffe57e08d870fd1b49a9e8b42212f89&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty