Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl chlorophosphate ≥97.0%

Supplier: Decon Laboratories Ltd

D2206-250G D2206-25G
TCD2206-250GEA 286 USD InStock
TCD2206-250G TCD2206-025G
Diethyl chlorophosphate ≥97.0%
Diethyl chlorophosphate
CAS Number: 814-49-3
MDL Number: MFCD00009075
Molecular Formula: C4H10ClO3P
Molecular Weight: 172.54
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 61
Specific Gravity (20/20): 1.20

Other Product Names / Synonyms: Chlorophosphoric Acid Diethyl Ester, Diethyl Phosphoryl Chloride


Formula: C₄H₁₀ClO₃P
MW: 172.55 g/mol
Boiling Pt: 93 °C
Density: 1.20 (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00009075
CAS Number: 814-49-3
EINECS: 212-396-4
UN: 3278
ADR: 6.1 I
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3a71b91f3fa5f3bc162607bd03abf2d5fa46b4e&vid=c3a71b91f3fa5f3bc162607bd03abf2d5fa46b4e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty