Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethoxybenzylamine ≥98.0%

Supplier: Decon Laboratories Ltd

D3494-25G D3494-5G
TCD3494-25GEA 138 USD InStock
TCD3494-25G TCD3494-5G
2,4-Dimethoxybenzylamine ≥98.0%
2,4-Dimethoxybenzylamine
CAS Number: 20781-20-8
MDL Number: MFCD00052393
Molecular Formula: C9H13NO2
Molecular Weight: 167.21
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 143
Flash Point (°C): 113
Specific Gravity (20/20): 1.11


Formula: C₉H₁₃NO₂
Boiling Pt: 143 °C
Density: 1.11 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00052393
CAS Number: 20781-20-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80c6680f1c553ef84b0c0307f172408e280a1f2a&vid=80c6680f1c553ef84b0c0307f172408e280a1f2a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty