Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl hexafluoroglutarate ≥96.0%

Supplier: Decon Laboratories Ltd

D3589-1G D3589-5G
TCD3589-1GEA 31 USD InStock
TCD3589-1G TCD3589-5G
Dimethyl hexafluoroglutarate ≥96.0%
Dimethyl hexafluoroglutarate
CAS Number: 1513-62-8
MDL Number: MFCD00155851
Molecular Formula: C7H6F6O4
Molecular Weight: 268.11
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Melting point (°C): -32
Flash Point (°C): 93
Specific Gravity (20/20): 1.50

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Dimethyl Perfluoroglutarate, Dimethyl Hexafluoropentanedioate, Hexafluoroglutaric Acid Dimethyl Ester, Perfluoroglutaric Acid Dimethyl Ester, Hexafluoropentanedioic Acid Dimethyl Ester


Formula: C₇H₆F₆O₄
Boiling Pt: 100 °C
Melting Pt: –32 °C
Density: 1.50 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00155851
CAS Number: 1513-62-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=955c25037a084e87b90fefb80607bb1da9006110&vid=955c25037a084e87b90fefb80607bb1da9006110&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty