Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dicyclohexylimidazolium tetrafluoroborate ≥98.0%

Supplier: Decon Laboratories Ltd

D3850-5G D3850-1G D3850-25G
TCD3850-5GEA 300 USD InStock
TCD3850-5G TCD3850-1G TCD3850-25G
1,3-Dicyclohexylimidazolium tetrafluoroborate ≥98.0%
1,3-Dicyclohexylimidazolium tetrafluoroborate
CAS Number: 286014-38-8
MDL Number: MFCD04973310
Molecular Formula: C15H25BF4N2
Molecular Weight: 320.18
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 174


Formula: C₁₅H₂₅BF₄N₂
MW: 320.18 g/mol
Melting Pt: 174 °C
MDL Number: MFCD04973310
CAS Number: 286014-38-8
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty