Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethylbutyric acid ≥98.0%

Supplier: Decon Laboratories Ltd

E0070-500ML E0070-25ML
TCE0070-500MLEA 132 USD InStock
TCE0070-500ML TCE0070-025ML
2-Ethylbutyric acid ≥98.0%
2-Ethylbutyric acid
CAS Number: 88-09-5
MDL Number: MFCD00002670
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 94
Freezing point (°C): -15
Specific Gravity (20/20): 0.92

Other Product Names / Synonyms: Diethylacetic Acid


Formula: C₆H₁₂O₂
Boiling Pt: 193 °C
Melting Pt: –14 °C
Density: 0.92 (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00002670
CAS Number: 88-09-5
EINECS: 201-796-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03137
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ab08c3f3578e74ca45e2e5e21e0b51dedacd1a&vid=47ab08c3f3578e74ca45e2e5e21e0b51dedacd1a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty