Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-1-hexene ≥95.0%

Supplier: Decon Laboratories Ltd

E0124-10ML E0124-25ML
TCE0124-010MLEA 167 USD InStock
TCE0124-010ML TCE0124-25ML
2-Ethyl-1-hexene ≥95.0%
2-Ethyl-1-hexene
CAS Number: 1632-16-2
MDL Number: MFCD00027242
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Specific Gravity (20/20): 0.73


Formula: C₈H₁₆
MDL Number: MFCD00027242
CAS Number: 1632-16-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1c95223474fde26eeb93b931b4db38f6390683d&vid=b1c95223474fde26eeb93b931b4db38f6390683d&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty