Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethylimidazole ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

E0132-250G E0132-25G
TCE0132-250GEA 192 USD InStock
TCE0132-250G TCE0132-025G
1-Ethylimidazole ≥98.0% (by GC, titration analysis)
1-Ethylimidazole
CAS Number: 7098-07-9
MDL Number: MFCD00055389
Molecular Formula: C5H8N2
Molecular Weight: 96.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 226
Specific Gravity (20/20): 1.00


Formula: C₅H₈N₂
MDL Number: MFCD00055389
CAS Number: 7098-07-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d41773bfc2d44fbda797dd1870b486534327ce&vid=b2d41773bfc2d44fbda797dd1870b486534327ce&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty