Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethylaniline ≥98.0%

Supplier: Decon Laboratories Ltd

E0364-25ML E0364-500ML
TCE0364-025MLEA 167.4 USD InStock
TCE0364-025ML TCE0364-500ML
3-Ethylaniline ≥98.0%
3-Ethylaniline
CAS Number: 587-02-0
MDL Number: MFCD00007818
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 214
Melting point (°C): -8
Flash Point (°C): 85
Specific Gravity (20/20): 0.97


Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 212
Melting Pt: -8
Density: 0.97 (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007818
CAS Number: 587-02-0
EINECS: 209-594-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e91478fe86f1ab9d8f0a155fef610d0a1eb9ac5&vid=5e91478fe86f1ab9d8f0a155fef610d0a1eb9ac5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty