Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluoranthene ≥98.0%

Supplier: Decon Laboratories Ltd
Warning

F0016-25G F0016-500G F0016-100G
TCF0016-025GEA 37.63 USD InStock
TCF0016-025G TCF0016-500G TCF0016-100G
Fluoranthene ≥98.0%
Fluoranthene
CAS Number: 206-44-0
MDL Number: MFCD00001184
Molecular Formula: C16H10
Molecular Weight: 202.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 384
Melting point (°C): 110
Flash Point (°C): 198


Formula: C₁₆H₁₀
Boiling Pt: 384 °C
Melting Pt: 110 °C
Flash Pt: 198 °C
MDL Number: MFCD00001184
CAS Number: 206-44-0
EINECS: 205-912-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13fe5c4620652109346ea3ad2cd1769dace7895c&vid=13fe5c4620652109346ea3ad2cd1769dace7895c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty