Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl fluoroacetate ≥98.0%

Supplier: Decon Laboratories Ltd

F0030-25ML F0030-500ML
TCF0030-025MLEA 42.72 USD InStock
TCF0030-025ML TCF0030-500ML
Ethyl fluoroacetate ≥98.0%
Ethyl fluoroacetate
CAS Number: 459-72-3
MDL Number: MFCD00000450
Molecular Formula: C4H7FO2
Molecular Weight: 106.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 30
Specific Gravity (20/20): 1.10

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Fluoroacetic Acid Ethyl Ester


Formula: C₄H₇FO₂
Boiling Pt: 117 °C
Density: 1.10 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00000450
CAS Number: 459-72-3
EINECS: 207-297-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a60f6b18b1a62b3e1358483c4370963ecd6bc70&vid=1a60f6b18b1a62b3e1358483c4370963ecd6bc70&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty